2-(Aminomethyl)phenol
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:932-30-9
:
Formula:C₇H₉NO
MDL Number:MFCD00870498
:123.1525 g/mol
Melting Pt:124…128 °C
EINECS:213-249-7
:GHS07
Storage Temperature:Ambient
Frequently Bought Together