Mithramycin A (from Streptomyces argillaceus)
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:18378-89-7
:
Formula:C₅₂H₇₆O₂₄
MDL Number:MFCD00135618
:1085.1 g/mol
Melting Pt:180…183 °C
:GHS07
Storage Temperature:Freezer
Frequently Bought Together