Face Shields, V5 series
Catalog # MMMA5F-11
Supplier: 3M
Specifications
- Barva:Bezbarvá
- Popis:V5 series faceshield
- D׊:244×462 mm
- Bal.:1
Specifications
About this item
Optical class 1, clear curved polycarbonate faceshield with anti-fog and anti-scratch coating. Compatible with 3M™ V5 Faceshield System for mounting on 3M™ Hard Hat Series G3000 or G22.
- Provides protection against secondary welding light
- Anti-fog and anti-scratch coated
- Snap in feature makes it quick and easy to connect
- Medium energy impact, Grade B
- Optical class 1 visor, low energy impact
Get protection against impacts and secondary welding light with 3M™ Faceshield, 5F-11. It provides impact protection B against medium speed particles (0.86 g steel ball at 120 m/s), UV, or intense visible light. Anti-fog and anti-scratch coated, with a curved shape that allows for a close fit to the face.
Designed to fit 3M™ Hard Hat Series G3000 or G22 with 3M™ V5 Faceshield Mounting system. The snap in feature makes it quick and easy for the faceshield to connect to the Faceshield Holder.
Certifications: EN 166:2001