tert-Butyl 3-(trifluoromethyl)-1,4,5,7-tetrahydro-6H-pyrazolo[3,4-c]pyridine-6-carboxylate
Catalog # APOSPC48634-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Typ balení:Bottle
- Bal.:5 g
- Sekvence:3173
Specifications
About this item
:
:291.27 g/mol
Storage Temperature:Ambient
CAS Number:733757-89-6
Formula:C₁₂H₁₆F₃N₃O₂
MDL Number:MFCD11044974