Order Entry
Czech Republic
ContactUsLinkComponent
 
[4-(Difluoromethyl)-6-(1,3-dimethyl-1H-pyrazol-4-yl)-3-methyl-1H-pyrazolo[3,4-b]pyridin-1-yl]acetic acid
Catalog # APOSPC410447-1G
Supplier:  Apollo Scientific
CAS Number:  
[4-(Difluoromethyl)-6-(1,3-dimethyl-1H-pyrazol-4-yl)-3-methyl-1H-pyrazolo[3,4-b]pyridin-1-yl]acetic acid
Catalog # APOSPC410447-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Typ balení:
    Bottle
  • Bal.:
    1 g
  • Sekvence:
    3047

Specifications

About this item

 :
 :335.309 g/mol
Storage Temperature:Ambient
CAS Number:1006444-98-9
Formula:C₁₅H₁₅F₂N₅O₂
MDL Number:MFCD08697729