Order Entry
Czech Republic
ContactUsLinkComponent
2-({4-Ethyl-5-[1-methyl-3-(trifluoromethyl)-1H-pyrazol-5-yl]-4H-1,2,4-triazol-3-yl}sulphanyl)acetohydrazide
2-({4-Ethyl-5-[1-methyl-3-(trifluoromethyl)-1H-pyrazol-5-yl]-4H-1,2,4-triazol-3-yl}sulphanyl)acetohydrazide
Catalog # APOSPC410365-1G
Supplier:  Apollo Scientific
CAS Number:  
2-({4-Ethyl-5-[1-methyl-3-(trifluoromethyl)-1H-pyrazol-5-yl]-4H-1,2,4-triazol-3-yl}sulphanyl)acetohydrazide
Catalog # APOSPC410365-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Typ balení:
    Bottle
  • Bal.:
    1 g
  • Sekvence:
    3047

Specifications

About this item

 :
 :349.335 g/mol
Storage Temperature:Ambient
CAS Number:1005700-45-7
Formula:C₁₁H₁₄F₃N₇OS
MDL Number:MFCD03421859