Order Entry
Czech Republic
ContactUsLinkComponent
2-[1-(2-Fluoro-benzyl)-piperidin-4-yl]-1H-benzoimidazole
2-[1-(2-Fluoro-benzyl)-piperidin-4-yl]-1H-benzoimidazole
Catalog # APOSPC408414-500MG
Supplier:  Apollo Scientific
CAS Number:  
2-[1-(2-Fluoro-benzyl)-piperidin-4-yl]-1H-benzoimidazole
Catalog # APOSPC408414-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Typ balení:
    Bottle
  • Bal.:
    500 mg
  • Sekvence:
    2725

Specifications

About this item

 :
 :309.381 g/mol
Storage Temperature:Ambient
CAS Number:887217-21-2
Formula:C₁₉H₂₀FN₃
MDL Number:MFCD05254852