Order Entry
Czech Republic
ContactUsLinkComponent
4-[(3-Chloro-5-trifluoromethyl-pyridin-2-yl)-methyl-amino]-piperidine-1-carboxylic acid tert-butyl ester
4-[(3-Chloro-5-trifluoromethyl-pyridin-2-yl)-methyl-amino]-piperidine-1-carboxylic acid tert-butyl ester
Catalog # APOSPC408403-500MG
Supplier:  Apollo Scientific
CAS Number:  
4-[(3-Chloro-5-trifluoromethyl-pyridin-2-yl)-methyl-amino]-piperidine-1-carboxylic acid tert-butyl ester
Catalog # APOSPC408403-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Typ balení:
    Bottle
  • Bal.:
    500 mg
  • Sekvence:
    2725

Specifications

About this item

 :
 :393.832 g/mol
Storage Temperature:Ambient
CAS Number:1417793-99-7
Formula:C₁₇H₂₃ClF₃N₃O₂
MDL Number:MFCD22631680