Order Entry
Czech Republic
ContactUsLinkComponent
(2-Nitro-5-trifluoromethylphenyl)acetic acid
(2-Nitro-5-trifluoromethylphenyl)acetic acid
Catalog # APOSPC407026-1G
Supplier:  Apollo Scientific
CAS Number:  
(2-Nitro-5-trifluoromethylphenyl)acetic acid
Catalog # APOSPC407026-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Typ balení:
    Bottle
  • Bal.:
    1 g
  • Sekvence:
    3047

Specifications

About this item

 :
 :249.143 g/mol
Storage Temperature:Ambient
CAS Number:927801-01-2
Formula:C₉H₆F₃NO₄
MDL Number:MFCD08436986