Order Entry
Czech Republic
ContactUsLinkComponent
3-(2-Chloro-6-fluorophenyl)-N-(2-{[(fur-2-yl)methyl]sulphanyl}ethyl)-5-methylisoxazole-4-carboxamide
Irritant
3-(2-Chloro-6-fluorophenyl)-N-(2-{[(fur-2-yl)methyl]sulphanyl}ethyl)-5-methylisoxazole-4-carboxamide
Catalog # APOSPC31870-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
3-(2-Chloro-6-fluorophenyl)-N-(2-{[(fur-2-yl)methyl]sulphanyl}ethyl)-5-methylisoxazole-4-carboxamide
Catalog # APOSPC31870-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Typ balení:
    Bottle
  • Bal.:
    250 mg
  • Sekvence:
    2590

Specifications

About this item

 :
 :394.848 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:680211-94-3
Formula:C₁₈H₁₆ClFN₂O₃S
MDL Number:MFCD01566569