Neutralisation unit, SIMVA, behrotest®
Catalog # BEHRB00217922
Supplier: BEHR
Specifications
- Popis:SIMVAC Neutralisation unit
- Bal.:1
Specifications
About this item
For behr digestion units (K-block & Inkjel devices), extraction systems for acid vapours are a necessity. We differentiate between a simple water jet pump / SIMVAC and the behrosog3 extraction system and neutraliser.
- SIMVAC can be used for digestion blocks with up to 8 sample positions
- Only requires a water connection to generate the required extraction performance – nothing else
- Easy, robust and price effective