Ladles, long handle, detectable, SteriPlast®
30 ml, +, Modrá
Katalogové číslo 331-0361
Dodavatel: BURKLE
Přejít na:
Product Details & Documents
The ladle is suitable for taking samples of liquids and viscous media as well as powders and granulates. The ladle has a very smooth bottom and rounded corners. This makes it possible to reach into any corner of a container or to take medium directly from the bottom of the container. A long handle with indentations for fingers ensures a firm hold and precise work.
- PS detectable
- Individually packaged and sterilised, for single use
- Reaches into every corner, handle length 300 mm
- Can be detected by colour scanners, metal detectors or X-rays
- Can be used as part of HACCP/IFS/BRC foreign object management
- Clean room manufactured (clean room class 7)
- Complies with EU food requirements and FDA regulations
The disposable ladle can be used in the pharmaceutical industry, chemistry and cosmetics and the food and animal feed industries.
Specifikace
- Sterilní:+
- Kapacita:30 ml
- Barva:Modrá