Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Copro-Duo Kit
Copro-Duo Kit (2×24) The complete kit for a clean and reliable coprology
Copro-Duo Kit (2×24) The complete kit for a clean and reliable coprology
Copro-Duo Kit (2×24) The complete kit for a clean and reliable coprology
Catalog # 720-1182
Supplier:
RAL
CAS Number:
Expand image
Copro-Duo Kit (2×24) The complete kit for a clean and reliable coprology
Catalog # 720-1182
Supplier:
RAL
CAS Number:
Specifications
Bal.:
1 KIT
Sekvence:
0075
Specifications
About this item
Product Details
:
:
1
Product Details