0
0,00 Kč
Order Entry
Přihlásit se
/
Registrovat
Products
Services
Digital Solutions
Applications
Support
About
Expresní objednávka
Stav objednávky
Váš účet
Košík
Czech Republic
Expresní objednávka
Stav objednávky
Váš účet
Košík
ContactUsLinkComponent
Domů
Gram-Hücker R Kit
Gram-Hücker R Kit To differentiate Gram-positive from Gram-negative Bacteria
Gram-Hücker R Kit To differentiate Gram-positive from Gram-negative Bacteria
Gram-Hücker R Kit To differentiate Gram-positive from Gram-negative Bacteria
Catalog # 720-0350
Supplier:
RAL
CAS Number:
Expand image
Gram-Hücker R Kit To differentiate Gram-positive from Gram-negative Bacteria
Catalog # 720-0350
Supplier:
RAL
CAS Number:
Specifications
Bal.:
1 KIT
Sekvence:
0075
Specifications
About this item
Product Details
Documentation
:
:
1
Product Details
Documentation