Alkaline mercuric potassium iodide solution TS Reagent acc. Chinese Pharmacopoeia (ChP)
Catalog # REGECPNESSR
Supplier: REAGECON
CAS Number:
Specifications
- Typ balení:Plastová láhev
- Bal.:500 mL
- Sekvence:2725
Specifications
About this item
:
UN:2922
ADR:8,II
:GHS05
:GHS06
:GHS08