Orders LinkContactUsLinkComponent
Humidity module
Humidity module
Dodavatel:   Testo
Humidity module
Dodavatel:  Testo
Certifikáty
Humidity module
2 Dostupných variant níže
Product Details & Documents
Hygroscopic materials always try to form a moisture equilibrium with the surrounding air. This principle is used to measure the moisture in material by measuring moisture equilibrium.
  • Slim design (Ø 4 mm) with remote electronics
Ideal for tight measuring points, based on the moisture equilibrium measured, testo 635 automatically shows material moisture in % in the display.
Material moisture calculation is available for the following materials:
–Concrete
– High-insulation bricks
– Solid bricks
– Hard wood
– Soft wood
– Gas concrete
– Particle board
– Calcium sulphate floating screed
– Cement floating screed
– Lime sand brick
Materials are easy to select from the menu. Using the 'Material' user profile, the material can be accessed directly at the touch at a button.
Additional material characteristics can be stored in testo 635-2 using ComSoft.
Informace o dodávce: Supplied with thin humidity probe (Ø 4 mm) with PTFE protective cap and adhesive modelling clay (four items), including fixed cable.
Product Family Options
Informace o produktu
  • Sekvence
    Rozsah
    Přesnost
    Rozlišení
    Dostupnost
    Price
  • TEST0636.2135
    Humidity probe including 4 attachable teflon protection caps
    Humidity probe including 4 attachable teflon protection caps
    01
    T: 0...+40 °C
    RH: 0 - 100 %
    T: ±0,2 °C;
    RH: ±2,0 (2 - 98) %
    RH: 0,1 %
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 13 040,00 Kč
    13 040,00 Kč
    1 * 1 KS
       13 040,00 Kč
  • 620-0217
    Humidity probe head, attachable to radio handle
    Humidity probe head, attachable to radio handle
    02
    T: −20…+70 °C
    RH: 0 - 100 %
    T: ±0,3 °C;
    RH: ±2,0 (2 - 98) %
    T: 0,1 °C;
    RH: 0,1 %
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 5 940,00 Kč
    5 940,00 Kč
    1 * 1 KS
       5 940,00 Kč