Shoe covers, BizTex® Microporous type PB[6]
PRTWST44WHR
: Portwest
- Barva:Bílá
- Bal.:200
These protective overshoes are designed for use in all work environments where shoes or floors need protecting. Elastic finish to top of shoe ensures a secure fit.
- Made of microporous polypropylene/polyethylene
: This fabric conforms to EN 14126 biological protection and EN 1149-1 anti-static protection, EN 13034 Type PB [6].
: Do not iron and wash.