Pestle for micro-tissue grinders with screw caps
Catalog # 432-5004
Supplier: DWK Life Sciences
Specifications
- Určeno pro:Wheaton micro-tissue grinder with screw caps
- Popis:Pestle
- Bal.:2
Specifications
About this item
Replacement pestle for WHEATON® micro tissue grinder with cap, nominal capacity 0,5 ml.
- Manufactured from borosilicate glass 3.3
- With PTFE pestle and screw cap
- 13-425 open top screw cap and PTFE faced silicone rubber liner
- Autoclavable