Orders LinkContactUsLinkComponent
Spinner flasks, disposable, Corning®
 :   Corning
Spinner flasks, disposable, Corning®
 :  Corning
Spinner flasks, disposable, Corning®
Spinner flasks, disposable, Corning®
The Corning® disposable spinner flask system comes ready-to-use with paddle and integrated magnet, eliminating the need for time-consuming assembly or cleaning and reassembly.
  • Paddle size and height is optimised for each vessel size
  • Integrated magnet provides smooth, even rotation at required speeds on slow-speed stirrers
  • Heat build-up in the vessel is reduced by means of a specially designed flange that raised the vessel off the stir-plate surface for the 125 and 500 ml flasks only
Molded from virgin polystyrene and gamma-irradiated, each spinner flask system assures a clean and sterile unit that is nonpyrogenic. No more concerns with detergent residues or contamination.
Made of USP Class VI and ISO 10993-18 polystyrene, the vessel is comparable to conventional glass spinner flasks for growth of suspension cell lines and any attachment-dependent cultures using microcarrier beads.
Product Family Options
  • Sekvence
    Popis
    Kapacita
    Hrdlo
    Počet postranních ramen
    Doporučený pracovní objem
    Ø×H
  • 01
    Flasks with two solid caps on the side arms
    125 ml
    centre neck 70 mm Ø, side arm neck thread GL 25
    2
    30-75 ml
    63,5×145 mm
    1 * 12 KS
    1 * 12 KS - 32 820,00 Kč
    32 820,00 Kč
    1 * 12 KS
       32 820,00 Kč
  • 02
    Flasks with two solid caps on the side arms
    500 ml
    centre neck 100 mm Ø, side arm neck thread GL 45
    2
    100-300 ml
    87,3× 203,2 mm
    1 * 12 KS
    1 * 12 KS - 29 230,00 Kč
    29 230,00 Kč
    1 * 12 KS
       29 230,00 Kč
  • 03
    Flasks with two vented caps on the side arms
    500 ml
    centre neck 100 mm Ø, side arm neck thread GL 45
    2
    100-300 ml
    87,3× 203,2 mm
    1 * 12 KS
    1 * 12 KS - 33 550,00 Kč
    33 550,00 Kč
    1 * 12 KS
       33 550,00 Kč
  • 04
    Flasks with two solid caps on the side arms
    1000 ml
    side arm necks thread GL 45
    2
    300-1000 ml
    137×245 mm
    1 * 6 KS
    1 * 6 KS - 34 630,00 Kč
    34 630,00 Kč
    1 * 6 KS
       34 630,00 Kč
  • 05
    Flasks with two vented caps on the side arms
    1000 ml
    side arm necks thread GL 45
    2
    300-1000 ml
    137×245 mm
    1 * 6 KS
    1 * 6 KS - 34 630,00 Kč
    34 630,00 Kč
    1 * 6 KS
       34 630,00 Kč
  • 06
    Flasks with two solid caps on the side arms
    3000 ml
    side arm necks thread GL 45
    2
    1000-3000 ml
    188×259 mm
    1 * 4 KS
    1 * 4 KS - 28 020,00 Kč
    28 020,00 Kč
    1 * 4 KS
       28 020,00 Kč
  • 07
    Flasks with two vented caps on the side arms
    3000 ml
    side arm necks thread GL 45
    2
    1000-3000 ml
    188×259 mm
    1 * 4 KS
    1 * 4 KS - 28 020,00 Kč
    28 020,00 Kč
    1 * 4 KS
       28 020,00 Kč
Accessories
  •  
    Sekvence
    Popis
  • CORN401901
    21
    Modified impeller assembly for magnetically-driven 36 L Corning® Proculture® spinner flask, reusable
    1 * 1 KS - 33 310,00 Kč
  •  
    Sekvence
    Popis
    Závit
  • 734-4102
    07
    Vent cap, 0,2 μm vent, for 500, 1000 and 3000 ml spinner flasks
    GL45
    1 * 4 KS - 1 130,00 Kč
  •  
    Sekvence
    Popis
    Závit
  • 734-4101
    04
    Side arm aseptic transfer cap, dip tube with 0,2 μm vent, male luer lock, for 500 ml spinner flask
    GL45
    1 * 2 KS - 11 210,00 Kč
  •  
    Sekvence
    Popis
  • CORN3568
    36
    Flat cap, PE, disposable, orange, for 500, 1000 and 3000 ml spinner flasks
    1 * 4 KS - 1 030,00 Kč
  •  
    Sekvence
    Popis
    Závit
  • 734-4100
    06
    Side arm aseptic transfer cap, dip tube with 0,2 μm vent, male luer lock, for 3000 ml spinner flask
    GL45
    1 * 2 KS - 11 210,00 Kč
  •  
    Sekvence
    Popis
    Závit
  • 734-4098
    05
    Side arm aseptic transfer cap, dip tube with 0,2 μm vent, male luer lock, for 1000 ml spinner flask
    GL45
    1 * 2 KS - 11 210,00 Kč