Orders LinkContactUsLinkComponent
Fall protection harnesses, 2 point Plus
Fall protection harnesses, 2 point Plus
Harness plus, 1,5 kg, 2-point
Catalog # PRTWFP10RER
Supplier:   Portwest
Fall protection harnesses, 2 point Plus
Catalog # PRTWFP10RER
Supplier:  Portwest
Fall protection harnesses, 2 point Plus
Fall protection harnesses, 2 point Plus
Harness plus, 1,5 kg, 2-point
Přejít na:
Price
2 010,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Fall Protection is put in place to prevent the risks associated with falling from heights, reducing impact force, restricting obstacle/ground collision and restricting users from fall hazard areas.
  • Ergonomic, lightweight
  • Front and rear D-ring
  • Durable webbing with high breaking strength
  • Flat bar buckles
Features two chest DRings and flat bar buckles that are easy to use and fail safe. The chest and leg adjusters offer excellent fit and user comfort.
Made from nylon webbing, alloy steel, heat treated. Inspection cover for protection of product label. Retail box which aids presentation for retail sales.
Certifications: EN 361
Specifikace
  • Připojovací body:
    2-point
  • Popis:
    Harness plus
  • Hmotnost:
    1,5 kg