ContactUsLinkComponent
Browguard with mesh visor, PW93
Browguard with mesh visor, PW93
Catalog # PRTWPW93BKR
Supplier:  Portwest
undefined
Browguard with mesh visor, PW93
Catalog # PRTWPW93BKR
Supplier:  Portwest
Specifications
  • Popis:
    Galvanised carbon steel, PP browguard with mesh visor
  • D׊:
    70×47 mm
  • Model:
    PW93
  • Bal.:
    1
Specifications
About this item
Mesh visor with PP browguard, suitable for protection against large highspeed particles. The ratchet adjustment system is designed for easy use.
  • Wheel ratchet size adjustment for easy fitting
  • Soft foam sweat band included
  • Can be adapted to be used over prescription glasses
  • Individually packed for vending machines
Materials: galvanised carbon steel, polypropylene.
Certifications: EN 1731 (F); EN 166 (B); CE