General purpose gloves, fully dipped nitrile safety cuff, A302
Supplier: Portwest
Certificates
About this item
This extra durable heavyweight nitrile glove is fully dipped, giving even more protection against cuts, scrapes, punctures and tears. The safety cuff feature allows for these gloves to be removed instantly with ease.
- A wide and increasing range of hand protection styles is available
- Made up of cotton and nitrile
- Smooth heavy nitrile coating
- Canvas safety cuff for added protection
- Fully dipped thumb for increased coverage
Certifications: AS/NZS 2161 .2, AS/NZS 2161 .3, EN 388:2016 - 4.1.1.1.X, EN 420.
Specifications
- Catalog No:
- PRTWA302NARL
- PRTWA302NARXL
- Model:
- A302
- A302
- Obouruké/jednoruké:
- Jednoruké
- Jednoruké
- Jednorázové/znovupoužitelné:
- opakovatelně použitelné
- opakovatelně použitelné
- Typ manžety:
- Safety cuff
- Safety cuff
- Norma EN:
- EN 388:2016 - 4.1.1.1.X, EN 420
- EN 388:2016 - 4.1.1.1.X, EN 420