Combined Six Cation Standard II for IC
Catalog # HONCE2033-100ML
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Matrice:H₂O
- Typ balení:Plastic bottle
- Bal.:100 mL
- Popis:Combined Six Cation Standard II
Specifications
About this item
Certified Reference Material (CRM).
500 mg/L each of K+, Ca2+, 250 mg/L each of NH4+, Mg2+, 200 mg/L Na+, 50 mg/L Li+ in H₂O