Avantor® Hichrom HI-DAI 2, GC Columns
Dodavatel: Avantor
Certifikáty
O tomto produktu
Application specifc column for direct introduction of aqueous samples, thus minimizing sample preparation.
Max temperature limits may change depending on the film thickness.
Specifikace
- Catalog No:
- HI43-25-250-30
- HI43-32-300-30
- HI43-25-250-50
- HI43-32-300-50
- HI43-32-500-50
- HI43-32-500-30
- Phase name:
- HI-DAI 2
- HI-DAI 2
- HI-DAI 2
- HI-DAI 2
- HI-DAI 2
- HI-DAI 2
- Phase composition:
- Proprietary Phase developed for direct aqueous injections
- Proprietary Phase developed for direct aqueous injections
- Proprietary Phase developed for direct aqueous injections
- Proprietary Phase developed for direct aqueous injections
- Proprietary Phase developed for direct aqueous injections
- Proprietary Phase developed for direct aqueous injections
- Crossbond:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Teplotní limit:
- 320 °C
- 320 °C
- 320 °C
- 320 °C
- 320 °C
- 320 °C