Avantor® Hichrom HI-8 HT, GC Columns
Supplier: Avantor
Certificates
About this item
High Temperature general purpose column ideal for PCBs compounds, Pesticides, environmental analysis.
Max temperature limits may change depending on the film thickness.
Specifications
- Catalog No:
- HI88-53-010-30
- HI88-53-015-15
- HI88-53-010-10
- HI88-53-010-15
- HI88-53-025-15
- HI88-53-025-30
- HI88-53-015-30
- HI88-53-017-30
- HI88-25-010-30
- HI88-25-015-15
- HI88-25-010-15
- HI88-10-010-10
- HI88-10-010-5
- HI88-25-025-15
- HI88-25-025-30
- HI88-25-015-30
- HI88-32-010-30
- HI88-32-015-15
- HI88-32-010-10G2
- HI88-32-010-10
- HI88-32-010-15
- HI88-32-025-30
- HI88-32-015-30
- HI88-32-025-15
- Phase name:
- HI-8 HT
- HI-8 HT
- HI-8 HT
- HI-8 HT
- HI-8 HT
- HI-8 HT
- HI-8 HT
- HI-8 HT
- HI-8 HT
- HI-8 HT
- HI-8 HT
- HI-8 HT
- HI-8 HT
- HI-8 HT
- HI-8 HT
- HI-8 HT
- HI-8 HT
- HI-8 HT
- HI-8 HT
- HI-8 HT
- HI-8 HT
- HI-8 HT
- HI-8 HT
- HI-8 HT
- Phase composition:
- Proprietary Phase - high temperature
- Proprietary Phase - high temperature
- Proprietary Phase - high temperature
- Proprietary Phase - high temperature
- Proprietary Phase - high temperature
- Proprietary Phase - high temperature
- Proprietary Phase - high temperature
- Proprietary Phase - high temperature
- Proprietary Phase - high temperature
- Proprietary Phase - high temperature
- Proprietary Phase - high temperature
- Proprietary Phase - high temperature
- Proprietary Phase - high temperature
- Proprietary Phase - high temperature
- Proprietary Phase - high temperature
- Proprietary Phase - high temperature
- Proprietary Phase - high temperature
- Proprietary Phase - high temperature
- Proprietary Phase - high temperature
- Proprietary Phase - high temperature
- Proprietary Phase - high temperature
- Proprietary Phase - high temperature
- Proprietary Phase - high temperature
- Proprietary Phase - high temperature
- Polarity:
- Low-to-mid polarity stationary phase
- Low-to-mid polarity stationary phase
- Low-to-mid polarity stationary phase
- Low-to-mid polarity stationary phase
- Low-to-mid polarity stationary phase
- Low-to-mid polarity stationary phase
- Low-to-mid polarity stationary phase
- Low-to-mid polarity stationary phase
- Low-to-mid polarity stationary phase
- Low-to-mid polarity stationary phase
- Low-to-mid polarity stationary phase
- Low-to-mid polarity stationary phase
- Low-to-mid polarity stationary phase
- Low-to-mid polarity stationary phase
- Low-to-mid polarity stationary phase
- Low-to-mid polarity stationary phase
- Low-to-mid polarity stationary phase
- Low-to-mid polarity stationary phase
- Low-to-mid polarity stationary phase
- Low-to-mid polarity stationary phase
- Low-to-mid polarity stationary phase
- Low-to-mid polarity stationary phase
- Low-to-mid polarity stationary phase
- Low-to-mid polarity stationary phase
- Crossbond:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Teplotní limit:
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C