Avantor® Hichrom HI-50, GC Columns
Supplier: Avantor
Certificates
About this item
High polarity phase, ideal for detailed FAMEs and Dioxins isomers separation.
- Phase composition: 50% Cyanopropyl, 50% Methyl Polysiloxane
Max temperature limits may change depending on the film thickness.
Specifications
- Catalog No:
- HI35-32-020-105
- HI35-32-020-100
- HI35-20-010-105
- HI35-32-020-50
- HI35-20-010-100
- HI35-32-020-30
- HI35-20-010-50
- HI35-32-025-100
- HI35-20-010-40
- HI35-32-020-60
- HI35-20-020-100
- HI35-32-025-30
- HI35-20-010-60
- HI35-32-025-105
- HI35-20-020-40
- HI35-32-025-60
- HI35-32-025-50
- HI35-20-020-105
- HI35-20-020-60
- HI35-20-020-50
- HI35-18-010-10
- HI35-18-010-20
- HI35-25-020-100
- HI35-20-020-20
- HI35-25-020-30
- HI35-25-020-105
- HI35-25-020-60
- HI35-20-010-10
- HI35-20-010-20
- HI35-25-020-50
- HI35-25-025-105
- HI35-25-025-100
- HI35-25-025-50
- HI35-25-025-30
- HI35-25-025-60
- HI35-18-010-40
- Phase name:
- HI-50
- HI-50
- HI-50
- HI-50
- HI-50
- HI-50
- HI-50
- HI-50
- HI-50
- HI-50
- HI-50
- HI-50
- HI-50
- HI-50
- HI-50
- HI-50
- HI-50
- HI-50
- HI-50
- HI-50
- HI-50
- HI-50
- HI-50
- HI-50
- HI-50
- HI-50
- HI-50
- HI-50
- HI-50
- HI-50
- HI-50
- HI-50
- HI-50
- HI-50
- HI-50
- HI-50
- Phase composition:
- 50% Cyanopropyl, 50% Methyl Polysiloxane
- 50% Cyanopropyl, 50% Methyl Polysiloxane
- 50% Cyanopropyl, 50% Methyl Polysiloxane
- 50% Cyanopropyl, 50% Methyl Polysiloxane
- 50% Cyanopropyl, 50% Methyl Polysiloxane
- 50% Cyanopropyl, 50% Methyl Polysiloxane
- 50% Cyanopropyl, 50% Methyl Polysiloxane
- 50% Cyanopropyl, 50% Methyl Polysiloxane
- 50% Cyanopropyl, 50% Methyl Polysiloxane
- 50% Cyanopropyl, 50% Methyl Polysiloxane
- 50% Cyanopropyl, 50% Methyl Polysiloxane
- 50% Cyanopropyl, 50% Methyl Polysiloxane
- 50% Cyanopropyl, 50% Methyl Polysiloxane
- 50% Cyanopropyl, 50% Methyl Polysiloxane
- 50% Cyanopropyl, 50% Methyl Polysiloxane
- 50% Cyanopropyl, 50% Methyl Polysiloxane
- 50% Cyanopropyl, 50% Methyl Polysiloxane
- 50% Cyanopropyl, 50% Methyl Polysiloxane
- 50% Cyanopropyl, 50% Methyl Polysiloxane
- 50% Cyanopropyl, 50% Methyl Polysiloxane
- 50% Cyanopropyl, 50% Methyl Polysiloxane
- 50% Cyanopropyl, 50% Methyl Polysiloxane
- 50% Cyanopropyl, 50% Methyl Polysiloxane
- 50% Cyanopropyl, 50% Methyl Polysiloxane
- 50% Cyanopropyl, 50% Methyl Polysiloxane
- 50% Cyanopropyl, 50% Methyl Polysiloxane
- 50% Cyanopropyl, 50% Methyl Polysiloxane
- 50% Cyanopropyl, 50% Methyl Polysiloxane
- 50% Cyanopropyl, 50% Methyl Polysiloxane
- 50% Cyanopropyl, 50% Methyl Polysiloxane
- 50% Cyanopropyl, 50% Methyl Polysiloxane
- 50% Cyanopropyl, 50% Methyl Polysiloxane
- 50% Cyanopropyl, 50% Methyl Polysiloxane
- 50% Cyanopropyl, 50% Methyl Polysiloxane
- 50% Cyanopropyl, 50% Methyl Polysiloxane
- 50% Cyanopropyl, 50% Methyl Polysiloxane
- Polarity:
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- Crossbond:
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Teplotní limit:
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- Kód USP:
- G8
- G8
- G8
- G8
- G8
- G8
- G8
- G8
- G8
- G8
- G8
- G8
- G8
- G8
- G8
- G8
- G8
- G8
- G8
- G8
- G8
- G8
- G8
- G8
- G8
- G8
- G8
- G8
- G8
- G8
- G8
- G8
- G8
- G8
- G8
- G8