Orders LinkContactUsLinkComponent
Spill workfloor
Spill workfloor
Spill workfloor heavy for 4 drums, 1600×1600×150 mm
Katalogové číslo BRDY196262
Dodavatel:   Brady
Spill workfloor
Katalogové číslo BRDY196262
Dodavatel:  Brady
Spill workfloor
Spill workfloor
Spill workfloor heavy for 4 drums, 1600×1600×150 mm
Přejít na:
Price
24 260,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Workfloors are a great way to handle and store hazardous fluids and contain potentially hazardous leaks in storage and dispensing areas.
  • Can hold/carry drums or smaller containers
  • Can be easily assembled with other units by using the connector system
  • Compatible with most forklifts and pallet trucks
Specifikace
  • Popis:
    Spill workfloor heavy for 4 drums
  • Š×H×V:
    1600×1600×150 mm