Workstation lockout writer software app
Workstation lockout writer software app, BBP®31, BBP®33, BBP®85, i3300
Katalogové číslo BRDY145445
Dodavatel: Brady
Přejít na:
Product Details & Documents
Workstation lockout writer app allows you to create compliant lockout/tagout labels, tags and procedures in no time.
- It has printing templates, simple translations, easy-to-use image editing for callouts, and more
- Features drop down menus for navigation between hazards, energy sources and devices to help you create custom standards for your procedures
- Helps with OSHA standards compliance for the lockout of hazardous energy sources
- Includes import capabilities from Lockout-Pro® software
- Minimum 1 GB RAM
- 200 MB free desk space required
Operating system: Windows® 10;Windows® 7 (32- or 64-bit);Windows® 8 (32- or 64-bit);Windows® 8.1 (32- or 64-bit).
Specifikace
- Popis:Workstation lockout writer software app
- Určeno pro:BBP®31, BBP®33, BBP®85, i3300