Specifications
- Popis:Brady workstation sfety and facility identification suite on CD
- Bal.:1
- Určeno pro:BBP12, BBP30, BBP31 Sign and, BBP33, BBP35, BBP37, BBP85, BMP41, BMP51, BMP53, BMP61, BMP71, J2000, J5000, i3300, i5100, i7100, S3000, S3100, M611
Specifications
About this item
Keep your facility safe and compliant with the safety and facility identification suite. This group of apps makes it easy to create custom signs, labels and pipemarkers.
- Hardware: 1 gigahertz (GHZ) processor or faster
- RAM: 2GB
- Screen resolution: 1024×768 or higher (recommended), 800×600 high colour (minimum)
Design and print labels for gauges, gradient scales and more in support of visual workplace initiatives - compatible with BBP37 only.
This group of apps makes it easy to create custom signs, labels and custom designer, express signs, arc flash, arrow tap, pipemarker, lean visuals, template editer.
Applicable with Windows® 10;Windows® 7 (32- or 64-bit);Windows® 7 Enterprise;Windows® 7 Professional;Windows® 7 SPI;Windows® 7 Ultimate;Windows® 8 (32- or 64-bit);Windows® 8 Professional (32 or 64 bit);Windows® 8.1 (32- or 64-bit);Windows® 8.1 Enterprise;Windows® 8.1 Professional.
Languages available: Bulgarian, Chinese (Simplified), Chinese (Traditional), Croatian, Czech, Danish, Dutch, English (US), Estonian, Finnish, French (France), German, Hungarian, Italian, Japanese, Korean, Norwegian, Polish, Portuguese (Portugal), Romanian, Russian, Slovak, Slovenian, Spanish (Mexico), Spanish (Spain), Swedish, Turkish, Vietnamese.