Industrial gloves, HyFlex® 11-816
Supplier: Ansell
Certificates
About this item
This nylon/spandex liner with foam nitrile coating provides unparalleled comfort and extreme tactility.
- Ultra-thin ergonomic design provides extreme tactility to handle small parts easily
- Silicone-free: No transfer of silicone contaminants to metal parts prior to painting
- Skin-friendly: Proprietary washing process that efficiently extracts impurities to deliver a cleaner, more comfortable glove
- FORTIX™ Abrasion Resistance Technology delivers extended wear time for enhanced worker safety and productivity
The combination between the thin liner and coating provide extreme comfort and a barehand feel for workers who rely on fingertip sensitivity for precision tasks. The FORTIX™ coating lasts longer than comparable styles under abrasive conditions for a longer wear life. HyFlex® 11-816 also ensures a secure grip in dry environments.
Recommended for: Picking, fastening components; assembling and mounting components; fastening components and parts; general handling; transport & logistics.
Certifications: EN 388:2016 (4121A); Cat. II
REACH Compliant
REACH Compliant
Packaging: 12 pairs per bag.
Specifications
- Catalog No:
- ANSE11-816/8
- ANSE11-816/10
- ANSE11-816/11
- ANSE11-816/6
- ANSE11-816/7
- ANSE11-816/9
- Obouruké/jednoruké:
- Jednoruké
- Jednoruké
- Jednoruké
- Jednoruké
- Jednoruké
- Jednoruké
- Jednorázové/znovupoužitelné:
- opakovatelně použitelné
- opakovatelně použitelné
- opakovatelně použitelné
- opakovatelně použitelné
- opakovatelně použitelné
- opakovatelně použitelné
- Latex/bez latexu:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Pudrované/nepudrované:
- Nepudrované
- Nepudrované
- Nepudrované
- Nepudrované
- Nepudrované
- Nepudrované
- Materiál vložky:
- Nylon/Spandex
- Nylon/Spandex
- Nylon/Spandex
- Nylon/Spandex
- Nylon/Spandex
- Nylon/Spandex
- Provedení:
- Palm foam nitrile coated
- Palm foam nitrile coated
- Palm foam nitrile coated
- Palm foam nitrile coated
- Palm foam nitrile coated
- Palm foam nitrile coated
- Rozchod:
- 18
- 18
- 18
- 18
- 18
- 18
- Typ manžety:
- Pletené zápěstí
- Pletené zápěstí
- Pletené zápěstí
- Pletené zápěstí
- Pletené zápěstí
- Pletené zápěstí
- Norma EN:
- EN 388:2016 (4121A)
- EN 388:2016 (4121A)
- EN 388:2016 (4121A)
- EN 388:2016 (4121A)
- EN 388:2016 (4121A)
- EN 388:2016 (4121A)
- Antistatický:
- +
- +
- +
- +
- +
- +