α,α-Diethoxytoluene
Catalog # ABCRAB137067.0025
Supplier: abcr
CAS Number:
Specifications
- Typ balení:Glass bottle for solids
- Bal.:25 g
- Sekvence:3308
Specifications
About this item
:
EINECS:212-265-1
CAS Number:774-48-1
Formula:C₁₁H₁₆O₂
MDL Number:MFCD00038525