Titriplex® V (Kyselina diethylentriaminpentaoctová) analytical reagent, Supelco®
Catalog # 1.08426.0100
Supplier: Merck
CAS Number:
Specifications
- Typ balení:Plastová láhev
- Bal.:100 g
- Sekvence:3401
- Environmentally Preferable:
Specifications
Specification Test Results
- Solubility:4.8 g/l (25 °C)
- Melting Point:220 - 222 °C decomposes
- Molar Mass:393.34 g/mol
- Bulk Density:580 kg/m3
- Density:1.56 g/cm3
- pH:2 - 3 (H2O, 20 °C) (saturated solution)
Specification Test Results
About this item
Synonyms: N-Carboxymethyliminobis(ethylenenitrilo)tetra(acetic acid), Kestranal® Aza Pa, DTPA, Titriplex® V, Idranal V, Diethylenetriaminepentaacetic acid, 2,2',2'',2'''-((((Carboxymethyl)azanediyl)bis(ethane-2,1-diyl))bis(azanetriyl))tetraacetic acid, Diethylenetriamine-N,N,N',N'',N''-pentaacetic acid
:
Merck Index:13,07202
Melting Pt:220…222 °C
Density:1,558 g/cm³ (20 °C)
ADR:9,III
:M-Clarity™ quality level = MQ100
EINECS:200-652-8
UN:3077
:GHS08
:GHS07
Storage Temperature:Ambient
CAS Number:67-43-6
Formula:C₁₄H₂₃N₃O₁₀
MDL Number:MFCD00004289