
Chlorid-oxid zirkoničitý oktahydrát analytical reagent, Supelco®
Catalog # 1.08917.0100
Supplier: Merck
CAS Number:
Specifications
- Typ balení:Plastová láhev
- Bal.:100 g
- Sekvence:3401
Specifications
Specification Test Results
- Solubility:(20 °C) soluble
- Molar Mass:322.28 g/mol
- Bulk Density:800 kg/m3
- Density:1.91 g/cm3 (20 °C)
- pH:1 (50 g/l, H2O, 20 °C)
Specification Test Results
About this item
:
Melting Pt:> 150 °C
Density:1,91 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:3260
ADR:8,II
:M-Clarity™ quality level = MQ300
EINECS:231-717-9
:GHS05
CAS Number:13520-92-8
Formula:Cl₂OZr·8H₂O
MDL Number:MFCD00149898