Draslík 1000 mg/L v 0,5 M kyselině dusičné (from KNO₃) Certipur® standard pro AAS, Supelco®
Catalog # 1.70230.0100
Supplier: Merck
CAS Number:
Specifications
- Koncentrace:1000 mg/L
- Matrice:0,5 M HNO₃
- Typ balení:Plastic bottle
- Bal.:100 mL
- M-Clarity quality level:MQ100
- Nástavec:Draslík
- Symbol:K
Specifications
About this item
Certifications: Traceable to SRM from NIST.