Isohexane (mixture of isomers), LiChrosolv® for HPLC, Supelco®
Catalog # 1.04335.2500
Supplier: Merck
CAS Number:
Specifications
- Typ balení:Glass bottle
- Bal.:2,5 L
- Sekvence:5096
Specifications
About this item
:
Boiling Pt:60 °C (1013 hPa)
Melting Pt:–154 °C
Density:0,653 g/cm³ (25 °C)
UN:1208
ADR:3,II
:M-Clarity™ quality level = MQ100
Flash Pt:–33 °C
EINECS:295-570-2
REACH:01-2119484651-34
:GHS02
:GHS08
:GHS07
:GHS09
Storage Temperature:Ambient
CAS Number:92112-69-1
:73513-42-5
Formula:C₆H₁₄
MDL Number:MFCD00009406