Riboflavin, TECHNICAL
3308, Glass bottle, 25 g
Supplier: VWR Chemicals
Jump to:
Product Details & Documents
Synonyms: Vitamin B2, Lactoflavin, 7,8-Dimethyl-10-((2S,3S,4R)-2,3,4,5-tetrahydroxypentyl)benzo[g]pteridine-2,4(3H,10H)-dione, (-)-Riboflavin, Vitamine G
- CAS Number:83-88-5
- Formula:C₁₇H₂₀N₄O₆
- MDL Number:MFCD00005022
- Molecular Weight:376.37
- Merck Index:12,08367
- Melting Pt:280 °C
- Storage Temperature:Ambient
- EINECS:201-507-1
Documents
Certificates
Search by Lot NumberP20_VWRC27414.137_SPEC_EN.pdf spec_VWRC27414.137 spec_VWRC27414.137 spec_VWRC27414.137
Product specification
Specifications
Specification Test Results
- Identifikace:splňuje test
Specifications
- Typ balení:Glass bottle
- Bal.:25 g
- Sekvence:3308