Pump tubing for AKTA flux
Katalogové číslo 29-0609-48
Dodavatel: Cytiva
Specifikace
- Popis:Pump tubing, PFD, size 17
- Bal.:1
Specifikace
O tomto produktu
Pump tubing for AKTA flux, AKTA flux is a versatile cross flow filtration system for sample concentration and diafiltration (buffer exchange) as well as cell harvest and clarification.