USB camera, Moticam® Pro S5 Lite
Catalog # 630-3101
Supplier: MOTIC
Specifications
- Model:Moticam Pro S5 Lite
- Rozlišení:5 MP
- Typ sondy:sCMOS
- Sensor size:2/3"
- Snímková frekvence:2448×2048 @ 37,5 fps; 1224×1024 @ 88,4 fps, max.
- Podporovaný OS:Windows 7/8/10, MAC OSX 10.9, Linux or higher
- Připojení objektivu:C-Mount
- Zařízení podpůrné:TWAIN, SDK and DirectShow Driver
- Rozhraní:USB 3.1
- Snímač:sCMOS
- Software:Motic Images Plus 3.0 for Windows, OSX and Linux
- Bal.:1
Specifications
About this item
High Grade sCMOS USB Camera Moticam® Pro S5 Lite for microscopy for an ultimate imaging result for lowlight situations in Fluorescence and Polarization.
- Fast frame rates
- sCMOS sensors back-illuminated sensor technology
- USB 3.1 interface
- Ideal for fluorescence applications
- Motic Images Plus 3.0 software
Low Light, Fast Phenomena – Moticam Pro S5 Lite delivers a 5MP mid-size resolution for all biological/medical fields, showing a premium sensitivity for lowlight situations in Fluorescence and Polarization. The Global shutter data readout helps to accelerate data transfer from fast phenomena, from single-cell organisms to industrial work with clockworks or extrusion processes.
Ordering information: To be purchased separately – C-mount adapter not included to be purchased from the same brand of your microscope, compatible with your microscope model and fitting with the size of the chip of your Moticam.
Delivery: Includes CS ring adaptor, USB 3.1 cable, Motic 4-dot calibration slide, Motic Images Plus 3.0 for Windows/OSX/Linux.
Caution: Minimum computer requirements – 2 GHz dual-core, RAM memory 2 GB, video memory min. 512 MB.