Quartz microfibre filters for air sampling, grade QM-C, Whatman™
Dodavatel: Whatman products (Cytiva)
Certifikáty
O tomto produktu
QM-C 100% quartz microfiber filter paper features low trace metals and high temperature resistance. It is well-suited for heavy metal analysis of particulate matter and sampling of acidic gases, stacks, flues and aerosols in air monitors.
- 100% pure quartz microfiber, without any binder or organic chemicals
- Pre-fired filters eliminate firing step, saving time in prepartion process
- Air retention efficiency ≥99,95% at 0,3 μm DOP
QM-C high purity quartz microfiber filter addresses the challenge of heavy metal analysis in PM2.5/PM10. It is also suitable for organic chemical analyses such as PCBs and PAHs.
Certifications: Certified to conform to European and US EPA standards for heavy metal analysis in air particulate matter.
Specifikace
- Catalog No:
- WHAT1855-050
- WHAT1855-150
- WHAT1855-865
- WHAT1855-110
- WHAT1855-082
- WHAT1855-085
- WHAT1855-090
- WHAT1855-047
- WHAT1855-070
- WHAT1855-037
- WHAT1855-045
- WHAT1855-866
- Hmotnost:
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- Tloušťka:
- 475 μm
- 475 μm
- 475 μm
- 475 μm
- 475 μm
- 475 μm
- 475 μm
- 475 μm
- 475 μm
- 475 μm
- Průtok vzduchu:
- 19 s/100 ml/6,5 cm²
- 19 s/100 ml/6,5 cm²
- 19 s/100 ml/6,5 cm²
- 19 s/100 ml/6,5 cm²
- 19 s/100 ml/6,5 cm²
- 19 s/100 ml/6,5 cm²
- 19 s/100 ml/6,5 cm²
- 19 s/100 ml/6,5 cm²
- 19 s/100 ml/6,5 cm²
- 19 s/100 ml/6,5 cm²
- Maximální teplota:
- 1200 °C
- 1200 °C
- 1200 °C
- 1200 °C
- 1200 °C
- 1200 °C
- 1200 °C
- 1200 °C
- 1200 °C
- 1200 °C