Multi-element ICH Q3D and USP 232 kit for metals analysis, Specpure®
Catalog # 47384.6C
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Bal.:5
- Popis:Multi-Element ICH Q3D & USP 232 Kit for Metals Analysis
- Pk Info:5× 100 ml
Specifications
About this item
This kit includes the following five items:
47318-AE, Multi-Element ICH/USP Oral Standard Solution A
47370-AE, Multi-Element ICH/USP Oral Standard Solution B
47387-AE, Multi-Element ICH/USP Oral Standard Solution C
47325-AE, Multi-Element ICH/USP Oral Standard Solution D
47358-AE, Multi-Element Pharma Internal Standard Solution 1