Specifications
- Barva:Béžová
- Popis:Super 77™ Multi-purpose spray adhesive
- Bal.:500 mL
Specifications
About this item
This is an extremely versatile, fast-drying, and transparent spray adhesive that bonds a wide range of lightweight materials. The permanent bond provides fast results to keep projects moving ahead. The spray formula is easy to dispense and apply directly onto surfaces and materials where needed.
- The low soak-in of the adhesive eliminates clamping and dry time
- Repositionable while providing long-lasting strength
- Soft, non-dimpling glue lines
- Allows for a bond time of 15 seconds to 30 minutes
- Typical applications include general assembly, temporary holding, palletising, and label attachment
Aggressive tack provides a high initial grab and sufficient open time to position materials properly. High-solids content supports its low soak-in property, allowing time to position parts for best fit. This attribute also provides higher coverage than competing spray adhesives and helps create soft, non-dimpling glue lines. The combination of fast tack, strong bond, and clean appearance make it ideal for a wide variety of projects. Form strong bonds on a wide array of materials, including wood, cardboard, fabric, and cushion foam to plastic, metal, paper, and much more. Great for display assembly and other light industrial uses.