- Barva:Černá
- Popis:Reclosable fastener, modified acrylic adhesive
- Bal.:1 Roll
This reclosable fastener is the convenient alternative to traditional fastening methods, such as screws, nuts, or bolts. It is designed to deliver a durable, reclosable fastening solution (which can be opened and closed multiple times) that bonds to a variety of substrates including metals, powder-coated paints, and plastics such as acrylics, polycarbonate, and ABS.
- Snap-in-place installation replaces screws, nails, snaps, and rivets
- Closes with an audible snap
- Five times the tensile strength of typical hook-and-loop fasteners
- Moisture and UV resistant
- Helps reduce vibration
- Suitable for both indoor and outdoor use
This can be opened and closed over and over, up to 1000 times before losing 50% of the original tensile strength for repeated access or repositioning. The mushroom heads easily slide over each other allowing positioning of parts before they are snapped together to create a firm fastening attachment. When mated, the fasteners are hidden from view and provide an unbroken surface to help create a smooth, clean appearance. The Type 250 stem density polyolefin fastener mates to Type 170, 250, or 400 giving the user the option of different closure strength depending upon applications.