O tomto produktu
Traditional pH indicator test papers, supplied on a 4,8 metre roll, consisting of high quality filter paper impregnated with indicator and mixed indicator solutions then dried and cut to size.
- Roll format are easy to use and most common ranges are available in a refill format
- Quality assurance - tests are checked and calibrated against certified buffer solutions
- Wide and narrow pH ranges available
3 Dostupných variant níže
Product Details & Documents
Traditional pH indicator test papers, supplied on a 4,8 metre roll, consisting of high quality filter paper impregnated with indicator and mixed indicator solutions then dried and cut to size.
- Roll format are easy to use and most common ranges are available in a refill format
- Quality assurance - tests are checked and calibrated against certified buffer solutions
- Wide and narrow pH ranges available
Documents
Specifications for Product Family
Specifikace
- Catalog No:
- 1.09560.0003
- 1.09562.0003
- 1.09564.0003
- M-Clarity quality level:
- MQ100
- MQ100
- MQ100
Specifikace
Product Family Options
Informace o produktu
- pH rozsahpH stupniceDostupnostPrice
- 1.09560.0003Acilit®Specifikace:
More
Specifications
Cat. No.1.09560.0003M-Clarity quality levelMQ100 - 1.09562.0003Alkalit®Specifikace:
More
Specifications
Cat. No.1.09562.0003M-Clarity quality levelMQ100 - 1.09564.0003Neutralit®Specifikace:
More
Specifications
Cat. No.1.09564.0003M-Clarity quality levelMQ100