0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
4-(Dimethylamino)benzaldehyde
4-(Dimethylamino)benzaldehyd hydrochloric solution, Supelco®
4-(Dimethylamino)benzaldehyd hydrochloric solution, Supelco®
03891-250ML
:
Merck
:
4-(Dimethylamino)benzaldehyd hydrochloric solution, Supelco®
03891-250ML
:
Merck
:
Bal.:
250 mL
Sekvence:
2590
:
:
8,II
:
M-Clarity™ quality level = MQ100
:
1789
:
GHS05
:
GHS07
:
100-10-7
:
(CH₃)₂NC₆H₄CHO
:
MFCD00003381