Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Filtry
Předkolony a frity
Avantor® Hichrom, PEEK Frit
Avantor® Hichrom, PEEK Frit
Avantor® Hichrom, PEEK Frit
Catalog # HICHHI-1140
Supplier:
Avantor
Expand image
Avantor® Hichrom, PEEK Frit
Catalog # HICHHI-1140
Supplier:
Avantor
Specifications
Materiál frity:
PEEK
Porozita:
2,0 µm
Popis:
Frit
Bal.:
1
Specifications
About this item
Product Details
Product Details