Laboratory coats, SMS, with zip closure
Catalog # HOIT114520
Supplier: HUBEI ORIENT INTL TRADING
Specifications
- Velikost oblečení:XL
- Typ uzávěru:Se zipem
- Aplikace oblečení:Laboratory
- Rod:Unisex
- Popis:With zip closure
- Sekvence:03
- Bal.:50
- Hmotnost:60 g/m²
Specifications
About this item
These disposable laboratory coats made from SMS.
- With shirt, Korean, knitted collar
- Zip for closures
Weight: 25 to 55 g/m²
Colour: White/green/blue
Packaging: 5 pcs/ bag, 10 bags/case.