NANOCOLOR® Photometric Tube Tests Kits for Ammonium
Supplier: MACHEREY-NAGEL
Certificates
About this item
Photometric test kits for the determination of ammonium in water samples.
- Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
- Bar code cuvette identification for easy test call up in NANOCOLOR® spectrophotometers
- Precisely pre-dosed reagents with constant high quality from batch to batch
- Colored pictogram instructions and easy handling of the tubes
- 16 mm (OD) tubes for easy pipetting
The test kits are evaluated in 16 mm (AD) round cuvettes. They are pre-programmed on NANOCOLOR® photometers and are also compatible with METTLER TOLEDO Easy VIS and Easy UV. Programming data for other spectrophotometers are available.
Sea water analysis: Yes
Hazardous material: Yes
Remark: Measuring range on NANOCOLOR VIS II
Sludge reagent set: No
Digestion set: No
NanOx Metal: No
NanOx N: No
Delivery: Rugged box with 20 test tubes. Sufficient for 20 tests.
Specifications
- Model:MANA985003
- Plošina:NANOCOLOR® tube tests
- Skladovací teplota:15...25 °C
- Životnost police (měsíce):12
- Hmotnost:380 g (Including packaging)
Specifications
Related Information
Certificates
Search by Lot NumberAmmonium 10 Tube Test.pdf Ammonium 3 Tube Test .pdf NANOCOLOR® Photometric Tube Tests, Ammonium 200.pdf Photometric Tube Tests, NANOCOLOR® Ammonium 2000.pdf Photometric Tube Tests, NANOCOLOR® Ammonium 50.pdf
Additional Product Details
IFU Ammonium 10 Tube Test.pdf IFU Ammonium 100 Tube Test.pdf IFU Ammonium 3 Tube Test.pdf IFU NANOCOLOR® Photometric Tube Tests, Ammonium.pdf IFU Photometric tube tests, NANOCOLOR® ammonium 2000.pdf
More
MANA985006_TDS_EN
Datasheet
Frequently Bought Together