Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standardy
Standardy organické
Tetrandrine, for drug analysis, analytical standard, Supelco®
Tetrandrine, for drug analysis, analytical standard, Supelco®
Tetrandrine, for drug analysis, analytical standard, Supelco®
Catalog # T2695-1G
Supplier:
Merck
CAS Number:
Expand image
Tetrandrine, for drug analysis, analytical standard, Supelco®
Catalog # T2695-1G
Supplier:
Merck
CAS Number:
Specifications
Bal.:
1 g
Standard:
analytical standard
Čí. CAS:
518-34-3
Popis:
Tetrandrine
Popis:
Tetrandrine
Specifications
About this item
Product Details
Product Details