0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
Standardy
Standardy organické
2-Mercaptobenzimidazole, analytical standard, Supelco®
2-Mercaptobenzimidazole, analytical standard, Supelco®
Catalog # 51810-50MG
Supplier:
Merck
CAS Number:
Expand image
2-Mercaptobenzimidazole, analytical standard, Supelco®
Catalog # 51810-50MG
Supplier:
Merck
CAS Number:
Specifications
Bal.:
50 mg
Standard:
analytical standard
Čí. CAS:
583-39-1
Popis:
2-Mercaptobenzimidazole
Popis:
2-Mercaptobenzimidazole
Specifications
About this item
Product Details
Product Details