0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
Standardy
Standardy organické
5-Hydroxydiclofenac, analytical standard, Supelco®
5-Hydroxydiclofenac, analytical standard, Supelco®
Katalogové číslo 41694-25MG
Supplier:
Merck
CAS Number:
Expand image
5-Hydroxydiclofenac, analytical standard, Supelco®
Katalogové číslo 41694-25MG
Supplier:
Merck
CAS Number:
Specifications
Bal.:
25 mg
Standard:
analytical standard
Čí. CAS:
69002-84-2
Popis:
5-Hydroxydiclofenac
Popis:
5-Hydroxydiclofenac
Specifications
About this item
Product Details
Product Details